• USA Home
  • Search Results

  • 13diaminopropane741210976211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,3-Diaminopropane

2 Products
Synonym:   1,3-Propanediamine, DAP, TMEDA, Trimethylenediamine
Image of 1,3-Diaminopropane
CAS Number:
Linear Formula:
 NH2(CH2)3NH2
Molecular Weight:
 74.12
 Beilstein/REAXYS Number:
 605277
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Potency
Show More