• USA Home
  • Search Results

  • 13dibromobenzene2359010836111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,3-Dibromobenzene

1 Products
Image of 1,3-Dibromobenzene
CAS Number:
Empirical Formula (Hill Notation):
 C6H4Br2
Molecular Weight:
 235.90
 Beilstein/REAXYS Number:
 1904538
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More

Product #
Description
194395 1,3-Dibromobenzene, 97%,