• USA Home
  • Search Results

  • 13dicyanobenzene1281362617511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,3-Dicyanobenzene

1 Products
Synonym:   Isophthalodinitrile, Isophthalonitrile
Image of 1,3-Dicyanobenzene
CAS Number:
Linear Formula:
 C6H4(CN)2
Molecular Weight:
 128.13
 Beilstein/REAXYS Number:
 1072209
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
145858 1,3-Dicyanobenzene, 98%,