• USA Home
  • Search Results

  • 13diiminoisoindoline14516346811911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,3-Diiminoisoindoline

1 Products
Synonym:   1,3-Isoindolinediimine, Phthalimide diimide
Image of 1,3-<WBR>Diiminoisoindoline
CAS Number:
Empirical Formula (Hill Notation):
 C8H7N3
Molecular Weight:
 145.16
 Beilstein/REAXYS Number:
 118477
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More