• USA Home
  • Search Results

  • 13dimethyladamantane1642970279411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,3-Dimethyladamantane

1 Products
Synonym:   1,3-Dimethyltricyclo[3.3.1.13,7]decane
Image of 1,3-<WBR>Dimethyladamantane
CAS Number:
Empirical Formula (Hill Notation):
 C12H20
Molecular Weight:
 164.29
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More