• USA Home
  • Search Results

  • 13diphenyl13propanedione2242512046711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,3-Diphenyl-1,3-propanedione

2 Products
Synonym:   Dibenzoylmethane
Image of 1,3-<WBR>Diphenyl-<WBR>1,3-<WBR>propanedione
CAS Number:
Linear Formula:
 (C6H5CO)2CH2
Molecular Weight:
 224.25
 Beilstein/REAXYS Number:
 514910
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More