• USA Home
  • Search Results

  • 141013tetraoxa716diazacyclooctadecane262352397855411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,4,10,13-Tetraoxa-7,16-diazacyclooctadecane

1 Products
Synonym:   1,10-Diaza-18-crown-6
Image of 1,4,10,13-<WBR>Tetraoxa-<WBR>7,16-<WBR>diazacyclooctadecane
CAS Number:
Empirical Formula (Hill Notation):
 C12H26N2O4
Molecular Weight:
 262.35
 Beilstein/REAXYS Number:
 609764
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More