• USA Home
  • Search Results

  • 14bis4vinylphenoxybutane2943911230998511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,4-Bis(4-vinylphenoxy)butane

1 Products
Synonym:   JandaJel crosslinker
Image of 1,4-<WBR>Bis(4-<WBR>vinylphenoxy)<WBR>butane
CAS Number:
Empirical Formula (Hill Notation):
 C20H22O2
Molecular Weight:
 294.39
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More