• USA Home
  • Search Results

  • 14bisacryloylpiperazine19423634217211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,4-Bis(acryloyl)piperazine

1 Products
Synonym:   1,4-Di(acryloyl)piperazine, N,N′-Bis(acryloyl)piperazine, BAP, PDA, Piperazine diacrylamide
Image of 1,4-<WBR>Bis(acryloyl)<WBR>piperazine
CAS Number:
Empirical Formula (Hill Notation):
 C10H14N2O2
Molecular Weight:
 194.23
 Beilstein/REAXYS Number:
 745431
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More