• USA Home
  • Search Results

  • 14bisdicyclohexylphosphinobutane450666503836011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,4-Bis(dicyclohexylphosphino)butane

1 Products
Image of 1,4-<WBR>Bis(dicyclohexylphosphino)<WBR>butane
CAS Number:
Linear Formula:
 (C6H11)2P(CH2)4P(C6H11)2
Molecular Weight:
 450.66
 Beilstein/REAXYS Number:
 3368850
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Reaction Suitability
Show More