• USA Home
  • Search Results

  • 14bistrimethylsilylbutadiyne19442452607211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,4-Bis(trimethylsilyl)butadiyne

1 Products
Synonym:   1,4-Bis(trimethylsilyl)-1,3-butadiyne, BTMSBD
Image of 1,4-<WBR>Bis(trimethylsilyl)<WBR>butadiyne
CAS Number:
Linear Formula:
 (CH3)3SiC≡CC≡CSi(CH3)3
Molecular Weight:
 194.42
 Beilstein/REAXYS Number:
 1758268
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More