• USA Home
  • Search Results

  • 14butanediol901211063411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,4-Butanediol

3 Products
Synonym:   1,4-Butylene glycol, Tetramethylene glycol
Image of 1,4-Butanediol
CAS Number:
Linear Formula:
 HO(CH2)4OH
Molecular Weight:
 90.12
 Beilstein/REAXYS Number:
 1633445
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Potency
  Special Grade
Show More