• USA Home
  • Search Results

  • 14dichlorobenzene1470010646711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,4-Dichlorobenzene

3 Products
Image of 1,4-Dichlorobenzene
CAS Number:
Linear Formula:
 1,4-(Cl)2C6H4
Molecular Weight:
 147.00
 EC Index Number
 203-400-5
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Titertest
Show More