• USA Home
  • Search Results

  • 14dichlorobenzenesolution1470010646711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,4-Dichlorobenzene solution

1 Products
CAS Number:
Empirical Formula (Hill Notation):
 C6H4Cl2
Molecular Weight:
 147.00
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More