• USA Home
  • Search Results

  • 14dichlorobutane1270111056511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,4-Dichlorobutane

2 Products
Synonym:   Tetramethylene dichloride
Image of 1,4-Dichlorobutane
CAS Number:
Linear Formula:
 Cl(CH2)4Cl
Molecular Weight:
 127.01
 Beilstein/REAXYS Number:
 635673
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More