• USA Home
  • Search Results

  • 14diethynylbenzene1261593514811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,4-Diethynylbenzene

1 Products
Image of 1,4-Diethynylbenzene
CAS Number:
Linear Formula:
 HC≡CC6H4C≡CH
Molecular Weight:
 126.15
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
632090 1,4-Diethynylbenzene, 96%,