• USA Home
  • Search Results

  • 14difluorobenzene1140954036311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,4-Difluorobenzene

2 Products
Image of 1,4-Difluorobenzene
CAS Number:
Empirical Formula (Hill Notation):
 C6H4F2
Molecular Weight:
 114.09
 Beilstein Registry Number:
 1904541
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More