• USA Home
  • Search Results

  • 14dimethoxybenzene1381615078711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,4-Dimethoxybenzene

3 Products
Synonym:   p-Methoxyanisole, Dimethylhydroquinone, Hydroquinone dimethyl ether, NSC 7483
Image of 1,4-Dimethoxybenzene
CAS Number:
Linear Formula:
 C6H4(OCH3)2
Molecular Weight:
 138.16
 Beilstein/REAXYS Number:
 774605
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Assay
Show More