• USA Home
  • Search Results

  • 14dinitrobenzene1681110025411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,4-Dinitrobenzene

2 Products
Image of 1,4-Dinitrobenzene
CAS Number:
Linear Formula:
 C6H4(NO2)2
Molecular Weight:
 168.11
 Beilstein/REAXYS Number:
 1105828
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Anal. Standard Form
  Physical Form
  Special Grade
  Application(s)
Show More