• USA Home
  • Search Results

  • 14dioxane881112391111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,4-Dioxane

13 Products
Synonym:   Diethylene oxide, Dioxane
Image of 1,4-Dioxane
CAS Number:
Empirical Formula (Hill Notation):
 C4H8O2
Molecular Weight:
 88.11
 Beilstein/REAXYS Number:
 102551
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Anal. Standard Form
  Additive
  Quality clarifier
  Purity Grade
  Package quantity
  Color
  Refractive Index
  Agency/Method
  Restriction
  Vapor pressure
  Product Line
  Melting Point
  Explosion limit
  Density
  Autoignition temp
  Transmittance
  Transition temperature
  Potency
  UV-Absorption Values
  Application(s)
  Special Grade
  pH Value
  Physical Form
Show More