• USA Home
  • Search Results

  • 14dioxanesolution881112391111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,4-Dioxane solution

16 Products
Synonym:   1,4-Dioxane solution
Image of 1,4-Dioxane solution
CAS Number:
Empirical Formula (Hill Notation):
 C4H8O2
Molecular Weight:
 88.11
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Titertest
  Anal. Standard Form
  Chemical Composition
  Concentration
  Composite Dimension
Show More

Product #
Description
612073 1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 3 mm × 8 in.,
611905 1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 5 mm × 8 in.,
551368 1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D),
612111 1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), NMR tube size 3 mm × 8 in.,
631833 1,4-Dioxane solution, NMR reference standard, 5 mM in benzene-d6 (99.6 atom % D), NMR tube size 3 mm × 8 in.,
631868 1,4-Dioxane solution, NMR reference standard, 5 mM in chloroform-d (99.8 atom % D), NMR tube size 3 mm × 8 in.,
CRM48367 1,4-Dioxane solution, certified reference material, 2000 μg/mL in methanol, ampule of 1 mL,
684228 1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), NMR tube size 10 mm × 8 in.,
704806 1,4-Dioxane solution, NMR reference standard, 1 mM in D2O ("100", 99.96 atom % D), NMR tube size 5 mm × 7 in.,
704814 1,4-Dioxane solution, NMR reference standard, 10 mM in D2O ("100%", 99.96 atom % D), NMR tube size 5 mm × 7 in.,
704822 1,4-Dioxane solution, NMR reference standard, 0.1 mM in D2O ("100%", 99.96 atom % D), NMR tube size 5 mm × 7 in.,
736171 1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), NMR tube size 5 mm × 8 in.,
703354 1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 6.5 mm × 8 in.,
688355 1,4-Dioxane solution, NMR reference standard, 10 mM in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in.,
688371 1,4-Dioxane solution, NMR reference standard, 10 mM in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.,
612774 1,4-Dioxane solution, NMR reference standard, 10% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 5 mm × 8 in.,