• USA Home
  • Search Results

  • 14dithioerythritol15425689268811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,4-Dithioerythritol

4 Products
Synonym:   erythro-1,4-Dimercapto-2,3-butanediol, erythro-2,3-Dihydroxy-1,4-butanedithiol, Cleland’s reagent, DTE
Image of 1,4-Dithioerythritol
CAS Number:
Linear Formula:
 HSCH2CH(OH)CH(OH)CH2SH
Molecular Weight:
 154.25
 Beilstein Registry Number:
 1719756
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  pH Value
  UV-Absorption Values
  Melting Point
  Absorption
  Solubility
  Product Line
Show More