• USA Home
  • Search Results

  • 14phenylenediaminedihydrochloride1810662418011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,4-Phenylenediamine dihydrochloride

1 Products
Synonym:   1,4-Benzenediamine dihydrochloride
Image of 1,4-Phenylenediamine dihydrochloride
CAS Number:
Linear Formula:
 C6H4(NH2)2·2HCl
Molecular Weight:
 181.06
 Beilstein/REAXYS Number:
 3911749
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More