• USA Home
  • Search Results

  • 157triazabicyclo440dec5ene13920580714711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,5,7-Triazabicyclo[4.4.0]dec-5-ene

1 Products
Synonym:   1,3,4,6,7,8-Hexahydro-2H-pyrimido[1,2-a]pyrimidine, Hhpp, TBD
Image of 1,5,7-<WBR>Triazabicyclo[4.4.0]<WBR>dec-<WBR>5-<WBR>ene
CAS Number:
Empirical Formula (Hill Notation):
 C7H13N3
Molecular Weight:
 139.20
 Beilstein/REAXYS Number:
 3242
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Assay
Show More