• USA Home
  • Search Results

  • 15acetyldeoxynivalenol338358833796611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

15-Acetyldeoxynivalenol

1 Products
Synonym:   15-O-Acetyl-4-deoxynivalenol from Fusarium graminearum, 15-Acetoxy-3α,7α-dihydroxy-12,13-epoxytrichothec-9-en-8-one
Image of 15-<WBR>Acetyldeoxynivalenol
CAS Number:
Empirical Formula (Hill Notation):
 C17H22O7
Molecular Weight:
 338.35
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Special Grade
Show More