• USA Home
  • Search Results

  • 15anhydrodsorbitol1641615458511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,5-Anhydro-D-sorbitol

1 Products
Synonym:   1,5-Anhydro-D-glucitol
Image of 1,5-Anhydro-<SC>D</SC>-sorbitol
CAS Number:
Empirical Formula (Hill Notation):
 C6H12O5
Molecular Weight:
 164.16
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More