• USA Home
  • Search Results

  • 15crown5220263310027511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

15-Crown-5

1 Products
Synonym:   1,4,7,10,13-Pentaoxacyclopentadecane, 15C5
Image of 15-Crown-5
CAS Number:
Empirical Formula (Hill Notation):
 C10H20O5
Molecular Weight:
 220.26
 Beilstein/REAXYS Number:
 1618144
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More

Product #
Description
188832 15-Crown-5, 98%,