• USA Home
  • Search Results

  • 15diphenylcarbazide2422814022711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,5-Diphenylcarbazide

4 Products
Synonym:   1,5-Diphenylcarbazide
Image of 1,5-<WBR>Diphenylcarbazide
CAS Number:
Linear Formula:
 ((C6H5)NHNH)2CO
Molecular Weight:
 242.28
 EC Index Number
 205-403-7
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Suitability
  Melting Point
  Assay
Show More