• USA Home
  • Search Results

  • 15pentanediol1041511129511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,5-Pentanediol

4 Products
Synonym:   Pentamethylene glycol, 1,5-Pentanediol
Image of 1,5-Pentanediol
CAS Number:
Empirical Formula (Hill Notation):
 C5H12O2
Molecular Weight:
 104.15
 EC Index Number
 203-854-4
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
  Purity Grade
  Potency
Show More