• USA Home
  • Search Results

  • 1616dimethylprostaglandine2380523974625311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

16,16-Dimethylprostaglandin E2

1 Products
Synonym:   16,16-Dimethyl-PGE2
Image of 16,16-<WBR>Dimethylprostaglandin E<SUB>2</SUB>
CAS Number:
Empirical Formula (Hill Notation):
 C22H36O5
Molecular Weight:
 380.52
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Functional group
Show More