• USA Home
  • Search Results

  • 16dihydroxynaphthalene1601757544011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,6-Dihydroxynaphthalene

1 Products
Synonym:   1,6-Naphthalenediol
Image of 1,6-<WBR>Dihydroxynaphthalene
CAS Number:
Linear Formula:
 C10H6(OH)2
Molecular Weight:
 160.17
 Beilstein/REAXYS Number:
 1939032
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More