• USA Home
  • Search Results

  • 16hexanediol1181762911811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,6-Hexanediol

3 Products
Synonym:   Hexamethylene glycol
Image of 1,6-Hexanediol
CAS Number:
Linear Formula:
 HO(CH2)6OH
Molecular Weight:
 118.17
 EC Index Number
 211-074-0
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More