• USA Home
  • Search Results

  • 17aethynylestradiol296405763611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

17α-Ethynylestradiol

2 Products
Synonym:   17α-Ethynyl-1,3,5(10)-estratriene-3,17β-diol, 19-Nor-1,3,5(10),17α-pregnatrien-20-yne-3,17-diol, Ethinylestradiol
Image of 17α-Ethynylestradiol
CAS Number:
Empirical Formula (Hill Notation):
 C20H24O2
Molecular Weight:
 296.40
 Beilstein/REAXYS Number:
 2419975
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More