• USA Home
  • Search Results

  • 17amethyltestosterone302455818411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

17α-Methyltestosterone

2 Products
Synonym:   17α-Methyl-4-androsten-17β-ol-3-one, 17β-Hydroxy-17α-methyl-4-androsten-3-one, Mesterone, Methyltestosterone
Image of 17α-Methyltestosterone
CAS Number:
Empirical Formula (Hill Notation):
 C20H30O2
Molecular Weight:
 302.45
 Beilstein/REAXYS Number:
 2057425
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More