• USA Home
  • Search Results

  • 18aglycyrrhetinicacid47068144905411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

18α-Glycyrrhetinic acid

2 Products
Synonym:   18α-Glycyrrhetic acid, 18-Isoglycyrrhetinic acid, 3β-Hydroxy-11-oxo-18α,20β-olean-12-en-29-oic acid
Image of 18α-Glycyrrhetinic acid
CAS Number:
Empirical Formula (Hill Notation):
 C30H46O4
Molecular Weight:
 470.68
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More