• USA Home
  • Search Results

  • 18dihydroxy3methylanthraquinone2542448174311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,8-Dihydroxy-3-methylanthraquinone

1 Products
Synonym:   3-Methylchrysazin, Chrysophanic acid
Image of 1,8-<WBR>Dihydroxy-<WBR>3-<WBR>methylanthraquinone
CAS Number:
Empirical Formula (Hill Notation):
 C15H10O4
Molecular Weight:
 254.24
 Beilstein/REAXYS Number:
 1252300
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
Show More