• USA Home
  • Search Results

  • 1a25dihydroxyvitamind3416643222206311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1α,25-Dihydroxyvitamin D3

2 Products
Synonym:   1α,25-Dihydroxycholecalciferol, Calcitriol
Image of 1α,25-Dihydroxyvitamin D<SUB>3</SUB>
CAS Number:
Empirical Formula (Hill Notation):
 C27H44O3
Molecular Weight:
 416.64
 Beilstein/REAXYS Number:
 7559394
 EC Number:
 MDL number:
 NACRES:
 NA.79
 PubChem Substance ID:
 eCl@ss:
 34058003
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More