• USA Home
  • Search Results

  • 1a25dihydroxyvitamind361919d3solution4196512872316011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1α,25-Dihydroxyvitamin D3 (6,19,19-d3) solution

3 Products
Synonym:   1α,25-Dihydroxycholecalciferol (6,19,19-d3) solution, Calcitriol (6,19,19-d3) solution, Deuterated 1-α25-Dihydroxyvitamin D3 solution
Image of 1α,25-Dihydroxyvitamin D<SUB>3</SUB> (6,19,19-d<SUB>3</SUB>) solution
CAS Number:
Empirical Formula (Hill Notation):
 C27D3H41O3
Molecular Weight:
 419.65
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More