• USA Home
  • Search Results

  • 1aaatrifluoromtolylpiperazinehydrochloride266691601569311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-(α,α,α-Trifluoro-m-tolyl)piperazine hydrochloride

1 Products
Synonym:   N-[3-(Trifluoromethyl)phenyl]piperazine hydrochloride, TFMPP hydrochloride
Image of 1-(α,α,α-Trifluoro-<I>m</I>-tolyl)piperazine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C11H13F3N2 · HCl
Molecular Weight:
 266.69
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More