• USA Home
  • Search Results

  • 1acetonaphthone1702194198011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Acetonaphthone

1 Products
Synonym:   1-Acetylnaphthalene, Methyl 1-naphthyl ketone
Image of 1-Acetonaphthone
CAS Number:
Linear Formula:
 C10H7COCH3
Molecular Weight:
 170.21
 Beilstein/REAXYS Number:
 1100618
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More

Product #
Description
276766 1-Acetonaphthone, 97%,