• USA Home
  • Search Results

  • 1amino2propanol75117896611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-1-Amino-2-propanol

1 Products
Synonym:   (±)-Isopropanolamine
Image of (±)<WBR>-<WBR>1-<WBR>Amino-<WBR>2-<WBR>propanol
CAS Number:
Linear Formula:
 CH3CH(OH)CH2NH2
Molecular Weight:
 75.11
 Beilstein/REAXYS Number:
 605275
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More