• USA Home
  • Search Results

  • 1aza18crown6263333394115011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Aza-18-crown-6

1 Products
Synonym:   1,4,7,10,13-Pentaoxa-16-azacyclooctadecane
Image of 1-Aza-18-crown-6
CAS Number:
Empirical Formula (Hill Notation):
 C12H25NO5
Molecular Weight:
 263.33
 Beilstein/REAXYS Number:
 1074079
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More