• USA Home
  • Search Results

  • 1benzylimidazole15820423871511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Benzylimidazole

1 Products
Image of 1-Benzylimidazole
CAS Number:
Empirical Formula (Hill Notation):
 C10H10N2
Molecular Weight:
 158.20
 Beilstein/REAXYS Number:
 114571
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 39161001
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
116416 1-Benzylimidazole, 99%,