• USA Home
  • Search Results

  • 1bochexahydro14diazepine2002811227550011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Boc-hexahydro-1,4-diazepine

1 Products
Synonym:   1-Boc-homopiperazine, tert-Butyl homopiperazine-1-carboxylate
Image of 1-<WBR>Boc-<WBR>hexahydro-<WBR>1,4-<WBR>diazepine
CAS Number:
Empirical Formula (Hill Notation):
 C10H20N2O2
Molecular Weight:
 200.28
 Beilstein/REAXYS Number:
 7581789
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More