• USA Home
  • Search Results

  • 1bromo22methoxyethoxyethane183045414917611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Bromo-2-(2-methoxyethoxy)ethane

2 Products
Image of 1-<WBR>Bromo-<WBR>2-<WBR>(2-<WBR>methoxyethoxy)<WBR>ethane
CAS Number:
Linear Formula:
 CH3(OCH2CH2)2Br
Molecular Weight:
 183.04
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More