• USA Home
  • Search Results

  • 1bromo2ethylbenzene18506197322411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Bromo-2-ethylbenzene

1 Products
Image of 1-<WBR>Bromo-<WBR>2-<WBR>ethylbenzene
CAS Number:
Linear Formula:
 C2H5C6H4Br
Molecular Weight:
 185.06
 Beilstein/REAXYS Number:
 1929735
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More