• USA Home
  • Search Results

  • 1bromo2methyl1propene13500301769411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Bromo-2-methyl-1-propene

1 Products
Synonym:   Isocrotyl bromide
Image of 1-<WBR>Bromo-<WBR>2-<WBR>methyl-<WBR>1-<WBR>propene
CAS Number:
Linear Formula:
 (CH3)2C=CHBr
Molecular Weight:
 135.00
 Beilstein/REAXYS Number:
 1733844
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More