• USA Home
  • Search Results

  • 1bromo35difluorobenzene1929946196111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Bromo-3,5-difluorobenzene

1 Products
Image of 1-<WBR>Bromo-<WBR>3,5-<WBR>difluorobenzene
CAS Number:
Linear Formula:
 BrC6H3F2
Molecular Weight:
 192.99
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More