• USA Home
  • Search Results

  • 1bromododecane2492314315711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Bromododecane

2 Products
Synonym:   Dodecyl bromide, Lauryl bromide
Image of 1-Bromododecane
CAS Number:
Linear Formula:
 CH3(CH2)11Br
Molecular Weight:
 249.23
 Beilstein/REAXYS Number:
 506159
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Purity Grade
  Assay
Show More