• USA Home
  • Search Results

  • 1butanethiol901910979511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Butanethiol

4 Products
Synonym:   Butyl mercaptan
Image of 1-Butanethiol
CAS Number:
Linear Formula:
 CH3(CH2)3SH
Molecular Weight:
 90.19
 EC Index Number
 203-705-3
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Titertest
  Solubility
Show More